Methyl arachidate ≥98.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

A0900-25G A0900-5G
TCIAA0900-25GEA 358 EUR
TCIAA0900-25G TCIAA0900-5G
Methyl arachidate ≥98.0% (by GC)
Methyl arachidate
Formule: C₂₁H₄₂O₂
Kookpunt: 369°C
MDL Nummer: MFCD00009014
CAS nummer: 1120-28-1
Merck Index: 14,00764
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e9c204f7e2dc58125fc9425ec5ab678bbe115c94&vid=e9c204f7e2dc58125fc9425ec5ab678bbe115c94&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 45.0 to 50.0 deg-C
Solubility in hot Methanol almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR