4-(Methylthio)benzaldehyde

Leverancier: Apollo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

OR0776-100G OR0776-10G OR0776-50G
APOSOR0776-100GEA 57 EUR
APOSOR0776-100G APOSOR0776-10G APOSOR0776-50G
4-(Methylthio)benzaldehyde
4-(Methylthio)benzaldehyde
Formule: C₈H₈OS
Kookpunt: 267 °C (1013 hPa)
Dichtheid: 1,14 g/cm³ (20 °C)
MDL Nummer: MFCD00006948
CAS nummer: 3446-89-7
EINECS: 222-365-7
UN: 3334
ADR: 9,

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR