4-Methoxybenzyl mercaptan ≥98.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

M1165-25G M1165-5G
TCIAM1165-25GEA 158 EUR
TCIAM1165-25G TCIAM1165-5G
4-Methoxybenzyl mercaptan ≥98.0% (by GC)
4-Methoxybenzyl mercaptan
Formule: C₈H₁₀OS
Kookpunt: 126°C
Dichtheid: 1,11 g/cm³ (20 °C)
Bewaar temperatuur: Koelkast
MDL Nummer: MFCD00004871
CAS nummer: 6258-60-2
EINECS: 228-393-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4bc62e864cfb13a31f4eaa75743590f17099479f&vid=4bc62e864cfb13a31f4eaa75743590f17099479f&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR