Ethylamine 33% ∼6.7 mol/L in water
Leverancier: TCI
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Gevaar
|
Synoniemen:
Monoethylamine, Aminoethane, Ethanamine
E0055-25ML
E0055-500ML
TCIAE0055-25MLEA
17.6
EUR
TCIAE0055-25ML
TCIAE0055-500ML
Ethylamine 33% ∼6.7 mol/L in water
Ethylamine
Formule:
C₂H₇N MW: 45.08 g/mol Kookpunt: 17 Smeltpunt: -81.1 Dichtheid: 0.689 Vlampunt: -37 Bewaar temperatuur: Kamertemperatuur |
MDL Nummer:
MFCD00008160 CAS nummer: 75-04-7 EINECS: 200-834-7 UN: 2733 ADR: 3 II Merck Index: 14,03762 |
|
Specification Test Results
Appearance | Colorless to Almost colorless clear liquid |
Concentration (Neutralization titration) | 33.0 to 36.0 w/w% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...