Crucibles with lids, stainless steel
Leverancier: NICKEL ELECTRO
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
123163 124164 128167
NICK123163EA 36.6 EUR
NICK123163 NICK124164 NICK128167
Crucibles with lids, stainless steel
Kroezen
These stainless steel 304 crucibles with bright finish are much harder and last longer than porcelain types.
  • Non-magnetic
  • Shatterproof
Leveringsinformatie: Supplied with lids.