Draining racks
Leverancier: GLASWARENFABRIK KARL HECHT
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
49027010 49028010
HECH49027010EA 113 EUR
HECH49027010 HECH49028010
Draining racks
Rekken Afdruiprekken
Made of wire, plastic coated.
  • Good chemical resistance
  • Good temperature resistance
  • Easy to clean
Suitable for hanging or free standing test tubes and flasks with a plastic trough.