Flasks, Erlenmeyer, with standard ground joint
Leverancier: Kavalier
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
632426207940 632426207501 632426207252 632426207941 632426207500 632426207300 632426207102 632426207052 632426207950 632426207250
KAVA632426207940EA 70.8 EUR
KAVA632426207940 KAVA632426207501 KAVA632426207252 KAVA632426207941 KAVA632426207500 KAVA632426207300 KAVA632426207102 KAVA632426207052 KAVA632426207950 KAVA632426207250
Flasks, Erlenmeyer, with standard ground joint
Kolven Erlenmeyers
Made of borosilicate glass.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
Volume graduations and labelling area.