Flessen, ronde bodem, met standaard geslepen scharnierpunten
Leverancier: Kavalier
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
632426219958 632426219500 632426219940 632426219250_U 632426219250
KAVA632426219958EA 369 EUR
KAVA632426219958 KAVA632426219500 KAVA632426219940 KAVA632426219250_U KAVA632426219250
Flessen, ronde bodem, met standaard geslepen scharnierpunten
Kolven Rondbodemkolven
Borosilicate glass 3,3, clear.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
One centre neck, useful for distillation and extraction.