Goblet racks for liquid nitrogen freezers
Leverancier: TENAK
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
TE21922 TE21924 TE21926 TE21928
TENATE21922EA 99.48 EUR
TENATE21922 TENATE21924 TENATE21926 TENATE21928
Goblet racks for liquid nitrogen freezers
Rekken Rekken voor cryogene dozen
Goblet racks are used for cryogenic LN2 freezer and made of stainless steel.
Rack is made for standard 65 mm goblets with a height of 125 mm. The goblet holds between 50 and 70 straws. Two springs per shelf secure the goblet from slipping out.
The rack has a label holder for easy identification of the samples and a grab lifter, which makes it easier to grab the rack when wearing cryogloves.