Flasks, round bottom, with two or three necks and standard ground joints
Leverancier: Kavalier
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
632426223958 632426222950
KAVA632426223958EA 506 EUR
KAVA632426223958 KAVA632426222950
Flasks, round bottom, with two or three necks and standard ground joints
Kolven Rondbodemkolven
Borosilicate glass 3,3.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
Typical applications: distillation, extraction.