Stainless steel sieve cassettes, PULVERISETTE 25
Leverancier: FRITSCH
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
45.7440.10 45.7460.10 45.7500.10 45.7510.10 45.7520.10 45.7530.10
FRIT45.7440.10EA 572 EUR
FRIT45.7440.10 FRIT45.7460.10 FRIT45.7500.10 FRIT45.7510.10 FRIT45.7520.10 FRIT45.7530.10
Stainless steel sieve cassettes, PULVERISETTE 25
Zeven Zeven voor molens
Sieve cassettes determine the fineness of a sample. Sieve cassettes are provided with trapezoidal and square perforations.
Sieve cassettes with trapezoidal perforations improve size reduction through extra shearing action.
Sieve cassettes with square perforations are used for the grinding of brittle materials and materials of medium fineness with a narrow grain band.