Jars with screw cap
Leverancier: SEMADENI
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
5490 5491 5492 5496
SEMA5490EA 136 EUR
SEMA5490 SEMA5491 SEMA5492 SEMA5496
Jars with screw cap
Monsternamepotjes Containers voor algemeen gebruik
Jars made of PP, white,with screw caps made of red PP.
  • Very good chemical resistance
  • Very good temperature resistance
Suitable for powders, granules, pastes or cosmetic products. Physiologically harmless and suitable for food storage.