Aspirator bottles with screw cap and spigot
Leverancier: DELTALAB
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
19662 19664 19660
DELT19662EA 66.8 EUR
DELT19662 DELT19664 DELT19660
Aspirator bottles with screw cap and spigot
Flessen Doseerflessen
HDPE, translucent with screw cap and tightly fitting spigot.
  • Heavy duty bottles, wall thickness 2 mm
  • Can be used at temperatures below 0 °C
  • The spigot includes a seal ring to ensure a leakproof seal
Ideal for storing and dispensing liquids. Also recommended for use as settling bottles. Equipped with a carrying handle, the 25 litre sizes have two carrying handles.