Maatcilinders, hoge vorm, klasse B, borosilicaat Glas
Leverancier: Kavalier
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
632432151546 632432151130
KAVA632432151546EA 183 EUR
KAVA632432151546 KAVA632432151130
Maatcilinders, hoge vorm, klasse B, borosilicaat Glas
Cilinders Maatcilinders
Borosilicate glass 3.3, tall form, class B.
  • Hexagonal glass base, with spout
  • Amber graduations