Capillary columns, TraceGOLD™ TG-200MS
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
26084-1420 26084-1430 26084-2240 26084-2960 26084-2970
555-8896EA 823 EUR
555-8896 555-8895 555-8893 555-8890 555-8889
Capillary columns, TraceGOLD™ TG-200MS
Kolommen voor chromatografie
TraceGOLD™ TG-200MS GC columns are exceptionally inert mid-polarity columns with selectivity and elution order optimised for difficult separations.
  • Polar phase, Trifluoropropylmethyl polysiloxane solid phase resolves compounds that phenyl and cyano phases cannot
  • Outstanding thermal stability and low bleed
  • Suitable for use with sensitive detectors including ECDs, NPDs and MS
  • Equivalent to USP G6 phase
  • Confirmation column in combination with another GC column