Filter tips
Leverancier: DELTALAB
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
327-30 327-34R 327-19 327-26
DELT327-30EA 2410 EUR
DELT327-30 DELT327-34R DELT327-19 DELT327-26
Filter tips
Pipettips
These filter tips avoid aerosols and prevent pipette and / or sample contamination.
  • High transparency
  • Free of metals
  • Only racked tips: RNAse- and DNAse-free, pyrogen-free
  • Racks are made of autoclavable PP (up to 121 °C)