Fixed angle rotors, Fiberlite™, for use with Sorvall™ WX ultracentrifuges
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
096-067135 096-087056 096-247028 096-087051
521-4307EA 20600 EUR
521-4307 521-4310 521-4309 521-4308
Fixed angle rotors, Fiberlite™, for use with Sorvall™ WX ultracentrifuges
Centrifuge Rotoren
These lightweight carbon fibre rotors reduce run-times with higher G-forces.
  • Ensure structural integrity with corrosion- and fatigue-resistant design
  • Backed by a 15-year warranty
Fiberlite™ F37L-8 x 100
Compatible with: Thermo Scientific Sorvall™ WX Ultracentrifuge Series and Beckman™ Floor Model L Series (except Optima) centrifuges, Nalgene™ bottles and tubes
Recommended for: Separations of subcellular organelles or concentration of viruses.
Fiberlite™ F50L-24 x 1,5
Compatible with: Thermo Scientific Sorvall™ WX, Sorvall Discovery™ SE Series andMore Product Information
Order Now
Specificaties
Environmentally Preferable