Capillary columns, CP-Sil 8 CB for amines
Leverancier: VARIAN
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
CP7595 CP7596 CP7597 CP7595I5 CP7596I5 CP7598 CP7598I5 CP7599
561-7595EA 855 EUR
561-7595 VARICP7596 VARICP7597 VARICP7595I5 VARICP7596I5 VARICP7598 VARICP7598I5 VARICP7599
Capillary columns, CP-Sil 8 CB for amines
Kolommen voor chromatografie
CP-Sil 8 CB for amines is a base-deactivated 5% phenyl polydimethylsiloxane column that can be used for a wide range of amines.
  • Good inertness towards basic compounds for best accuracy
  • Guaranteed for the analysis of a broad range of amines for reliable results
  • Available in non-polar and polar phases for broad application range
Due to a thermal stability up to 350 °C, it analyzes a broad range of amines up to C20, as well as alkanolamines.