Capillary columns, CP-Wax for volatile amines and diamines
Leverancier: VARIAN
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
CP7422 CP7424
VARICP7422EA 787 EUR
VARICP7422 VARICP7424
Capillary columns, CP-Wax for volatile amines and diamines
Kolommen voor chromatografie
CP-Wax for volatile amines and diamines is optimised for analysis of volatile amines and diamines, particularly C3-C8 amines and diamines. The column delivers symmetrical peaks due to basic reagent deactivation and exhibits good inertness towards basic compounds.
  • Guaranteed analysis of volatile amines and diamines
  • Symmetrical peaks due to basic reagent deactivation
  • Good inertness towards basic compounds