Capillary columns, TRACE™ TR-FFAP
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
260N130P 260N142P 260N143P 260N154P 260N225P 260N230P 260N298P
555-9476EA 462 EUR
555-9476 555-9443 555-9411 555-9410 555-9407 555-9406 555-9405
Capillary columns, TRACE™ TR-FFAP
Kolommen voor chromatografie
TRACE™ TR-FFAP GC columns produce outstanding robustness in difficult-to-analyse separations.
  • Engineered specifically for FFAP analysis
  • Polar phase, TPA modified polyethylene glycol
  • Quality tested for acidic compound analysis
  • Equivalent to USP G25 and G35
  • Excellent quality and reproducibility
  • Low bleed even at elevated temperatures