Capillary columns, TRACE™ TR-V1
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
260V332P 260V333P 260V339P 260V341P 260V396P 260V495P
555-9436EA 837 EUR
555-9436 555-9435 555-9434 555-9433 555-9432 555-9428
Capillary columns, TRACE™ TR-V1
Kolommen voor chromatografie
Thick film columns for the analysis of volatile analytes.
  • Mid-polarity phase, 6% cyanopropylphenyl polysiloxane
  • Low bleed suitable for MS detection