HPLC columns, Hypersil GOLD™ Silica
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
25105-254630 25103-154630 25105-254030 25105-154630 25102-152130 25103-014001 25102-202130 25102-102130 25102-052130
554-4302EA 968 EUR
554-4302 554-4301 555-7590 555-7578 555-7754 554-4475 555-7755 555-7752 554-4300
HPLC columns, Hypersil GOLD™ Silica
Kolommen voor chromatografie
This unbonded, highly pure, base-deactivated silica media efficiently separates non polar and moderately polar organic compounds by normal phase chromatography.
  • Outstanding peak shape and sensitivity