Anion exchange bulk packing material
Leverancier: HAMILTON BONADUZ
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
79584 79585 79586 79594 79595 79596 79597 79598 79599 79827
553-0005EA 0 EUR
553-0005 553-0006 553-0007 553-0015 553-0016 553-0017 553-0018 553-0019 553-0020 553-0034
Anion exchange bulk packing material
Sorbenten voor chromatografie Sorbenten voor HPLC
Polymer bulk packing material for anion exchange applications.
Chromatographic capacity and efficiency tests are conducted on both bulk and packed column resins to ensure product integrity.