Sample bags, Whirl-Pak™
Leverancier: DELTALAB
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
200343 200347 200333
DELT200343EA 870 EUR
DELT200343 DELT200347 DELT200333
Sample bags, Whirl-Pak™
Zakken Monsterzakken
These LDPE blend sterile sample bags are resistant and transparent that are designed for both solid and liquid samples and the plastic thickness depends on the model. These are unique bags with a one-piece seam, avoiding the possible risks of the loss of the corners bags. To assure a leakproof closure to maintain the sample integrity, close the closing wires three times.
  • Ethylene oxide sterilised
  • Suitable for food industry
  • Non autoclavable
  • Leakproof closure with several metallic rounded sticks
  • The bags resist temperature up to 82 °C