Bottles, narrow neck, borosilicate glass, clear, with screw cap
Leverancier: DELTALAB
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
402001 402002 402005 402010
DELT402001EA 112 EUR
DELT402001 DELT402002 DELT402005 DELT402010
Bottles, narrow neck, borosilicate glass, clear, with screw cap
Flessen Glazen flessen
Bottles, made of borosilicate glass 3,3. Ideal for chemical reagents and culture media.
  • Autoclavable up to 140 °C
  • Graduations printed in white colour
  • Screw caps made of blue polypropylene
  • Ribbed screw caps for a better handling
  • Drip ring on bottle neck to avoid dripping