Sample dippers
Leverancier: DELTALAB
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
19575 19576
DELT19575EA 415 EUR
DELT19575 DELT19576
Sample dippers
Monsternemers Monsternemers voor vloeistoffen
The polypropylene cups have threaded fittings that screw onto the handle.
  • Autoclavable container up to 121 °C
  • Two pouring spouts for both left- or right- handed people
  • HDPE ribbed handles with hanging hook for a sure grip
Suitable for convenient sampling from tanks, vats, streams, ponds and lakes.
Order Now
Specificaties
Environmentally Preferable