Fine dosage syringes, Omnifix®-F Solo
Leverancier: B. Braun
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
9161406V 9204512
612-2899EA 29.7 EUR
612-2899 BRAU9204512
Fine dosage syringes, Omnifix®-F Solo
Spuiten Injectiespuiten, medisch
These fine dosage syringes are made of PP/PS. Plunger stopper with double sealing ring for slow aspiration and injection made of natural latex free synthetic material.
  • Highly transparent barrel with permanent marking
  • Safe plunger backstop
  • Easy aspiration up to maximum volume
  • Minimal residual volume
  • Latex free/PVC free
Certificaten: According to EN ISO 7886-1