(S)-(+)-Glycidyl methyl ether ≥97.0% (by GC)
Leverancier: TCI
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Gevaar
|
G0281-25G
G0281-5G
TCIAG0281-25GEA
371
EUR
TCIAG0281-25G
TCIAG0281-5G
(S)-(+)-Glycidyl methyl ether ≥97.0% (by GC)
(S)-(+)-Glycidyl methyl ether
Formule:
C₄H₈O₂ Vlampunt: 17°C |
MDL Nummer:
MFCD00273366 CAS nummer: 64491-68-5 UN: 1992 ADR: 3,II |
|
Specification Test Results
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 97.0 % |
Specific rotation [a]20/D | +12.0 to +15.0 deg (Neat) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...