N-(3,5-Dimethoxybenzyl)-2-(1,1,2,2,3,3,4,4,5,5,6,6,6-tridecafluorohexyl)benzamide

Leverancier: Thermo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

RF04575.250 RF04575.1
MAYBRF04575.250EA 0 EUR
MAYBRF04575.250 MAYBRF04575.1
N-(3,5-Dimethoxybenzyl)-2-(1,1,2,2,3,3,4,4,5,5,6,6,6-tridecafluorohexyl)benzamide
N-(3,5-Dimethoxybenzyl)-2-(1,1,2,2,3,3,4,4,5,5,6,6,6-tridecafluorohexyl)benzamide
Formule: C₂₂H₁₆F₁₃NO₃
MDL Nummer: MFCD00179696
CAS nummer: 288161-36-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR