(R)-(+)-2-Chlorobutyric acid ≥98.0% (by titrimetric analysis)
Leverancier: TCI
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Gevaar
|
C2109-1G
C2109-5G
TCIAC2109-1GEA
102
EUR
TCIAC2109-1G
TCIAC2109-5G
(R)-(+)-2-Chlorobutyric acid ≥98.0% (by titrimetric analysis)
(R)-(+)-2-Chlorobutyric acid
Formule:
C₄H₇ClO₂ Kookpunt: 98°C |
MDL Nummer:
MFCD00067098 CAS nummer: 54053-45-1 UN: 3265 ADR: 8,III |
|
Specification Test Results
Appearance | Colorless to Light yellow to Light orange clear liquid |
Purity (GC) | min. 96.0 % |
Optical purity (GC) | min. 96.0 ee% |
Purity (Neutralization titration) | min. 98.0 % |
Specific rotation [a]20/D | +15.0 to +19.0 deg (neat) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...