2-Chloro-4-fluorotoluene

Leverancier: Thermo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

SB01814.1 SB01814.10 SB01814.50
MAYBSB01814.1EA 0 EUR
MAYBSB01814.1 MAYBSB01814.10 MAYBSB01814.50
2-Chloro-4-fluorotoluene
2-Chloro-4-fluorotoluene
Formule: C₇H₆ClF
Kookpunt: 151,5…153 °C (1013 hPa)
Dichtheid: 1,1972 g/cm³ (20 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00000572
CAS nummer: 452-73-3
EINECS: 207-209-8
UN: 1993
ADR: 3,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR