4-Chloro-6-nitro-m-cresol ≥98.0% (by GC)
Leverancier: TCI
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Waarschuwing
|
C1684-25G
C1684-5G
TCIAC1684-25GEA
564
EUR
TCIAC1684-25G
TCIAC1684-5G
4-Chloro-6-nitro-m-cresol ≥98.0% (by GC)
4-Chloro-6-nitro-m-cresol
Formule:
C₇H₆ClNO₃ |
MDL Nummer:
MFCD00007114 CAS nummer: 7147-89-9 EINECS: 230-461-5 |
|
Specification Test Results
Appearance | Light yellow to Amber to Dark green powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 133.0 to 136.0 deg-C |
Solubility in Toluene | very faint turbidity |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...