5-Isoquinolineboronic acid ≥97%
Leverancier: Thermo Fisher Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Waarschuwing
|
H53230.MD
H53230.03
H53230.MDEA
61.2
EUR
H53230.MD
H53230.03
5-Isoquinolineboronic acid ≥97%
5-Isoquinolineboronic acid
Purity: 97%
CAS No.: 371766-08-4
Molecular Formula: C9H8BNO2
Formula Weight: 172.98
Melting Point: 204-206°
MDL No.: MFCD03839356
Formule:
C₉H₈BNO₂ Smeltpunt: 204…206 °C Bewaar temperatuur: Kamertemperatuur |
MDL Nummer:
MFCD03839356 CAS nummer: 371766-08-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...