Methyl-m-toluate
Leverancier: Thermo Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
TL00192.10 TL00192.50
MAYBTL00192.10EA 0 EUR
MAYBTL00192.10 MAYBTL00192.50
Methyl-m-toluate
Methyl-m-toluate
Formule: C₉H₁₀O₂
MDL Nummer: MFCD00008436
CAS nummer: 99-36-5
EINECS: 202-753-2