Sieve inserts for PULVERISETTE 15
Leverancier: FRITSCH
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
45.1200.10 45.1210.10 45.1220.10 45.1230.10 45.1240.10 45.1250.10 45.2200.10 45.2210.10
FRIT45.1200.10EA 184 EUR
FRIT45.1200.10 FRIT45.1210.10 FRIT45.1220.10 FRIT45.1230.10 FRIT45.1240.10 FRIT45.1250.10 FRIT45.2200.10 FRIT45.2210.10
Sieve inserts for PULVERISETTE 15
Zeven Zeven voor molens
Sieve inserts determine the final fineness of a sample. These sieve inserts are available with trapezoidal or square perforations.
Sieve inserts with trapezoidal perforations improve size reduction through extra shearing action.
Sieve inserts with square perforations are used for the grinding of brittle materials and materials of medium fineness with a narrow grain band.