HPLC columns, Acclaim™ RSLC 120 C8
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
072616 072617
555-7186EA 1060 EUR
555-7186 555-7187
HPLC columns, Acclaim™ RSLC 120 C8
Kolommen voor chromatografie
These reversed phase columns are used for the separation of analytes with moderate hydrophobicity. The unique chemistry of densely bonded C8 ligands on an ultrapure, spherical, porous silica substrate gives the excellent efficiencies.
  • Low silanol activity for excellent peak shapes
  • Reproducible column-to-column performance
  • Stable between pH 2 and 8
  • Extremely low bleed, fully compatible with MS
  • Compounds elute more quickly, yielding faster separations