HPLC columns, Acclaim™ RSLC Carbonyl
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
077972 077973 077974
555-7545EA 1140 EUR
555-7545 555-7546 555-7547
HPLC columns, Acclaim™ RSLC Carbonyl
Kolommen voor chromatografie
These silica based carbonyl columns provides ideal selectivity for baseline resolution and separation of DNPH derivatives of aldehydes and ketones.