HPLC columns, Acclaim™ PolarAdvantage
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
069691 069698 070079 071983 063693 061316 061317 061318 061319 061320 061321
555-7119EA 265 EUR
555-7119 555-7132 555-7167 555-7181 555-7300 555-7316 555-7317 555-7318 555-7319 555-7320 555-7321
HPLC columns, Acclaim™ PolarAdvantage
Kolommen voor chromatografie
These sulfonamide embedded columns are ideal for the separation of polar and non polar analytes. This provides an unique selectivity and excellent peak shape for nucleic acid, bases and pharmaceutical compounds using highly aqueous mobile phases.
  • Low silanol activity for excellent peak shape with basic compounds
  • High selectivity for hydrophobic aromatic molecules
  • Extremely low bleed, fully compatible with MS