Pipette tips, GripTips
Leverancier: Integra Biosciences
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
GripTips
613-3195EA 36.9 EUR
613-3195 613-3270 613-3206 613-3207 613-3216 613-3208 613-3264 613-3265 613-3196 613-3268 613-3202 613-3213 613-3203 613-3269 613-3211 613-3212 613-3267 613-3201
Pipette tips, GripTips
Pipettips
GripTips are manufactured from virgin medical grade materials and designed for use with the VIAFLO pipettes. Their features perfectly match the tip fitting to form a complete system. Without hammering, the GripTips literally snap onto the pipette's tip fitting, which prevents tips from falling off.
  • Reduced tip attachment and ejection forces for comfortable, stress-free pipetting
  • Extended length design enables easy access into tubes and deep well blocks
  • Sterile tips are certified free of RNase, DNase, pyrogens, PCR inhibitors and endotoxins
  • GREEN CHOICE refill system: environmentally friendly refill system to reuse the current tip racks and thus reduce plastic waste by 60%
Verpakking: Available in bulk packs, tip racks with single inserts in 96 and 384 fold configuration or as GREEN CHOICE refills.