Capillary columns, TraceGOLD™ TG-624
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
26085-3390 26085-3330 26085-3410 26085-3960 26085-4080 26085-4090 26085-4900 26085-4950 26085-4960 26085-3320
555-8886EA 901 EUR
555-8886 555-8887 555-8885 555-8884 555-8883 555-8882 555-8881 555-8880 555-8941 555-8888
Capillary columns, TraceGOLD™ TG-624
Kolommen voor chromatografie
These GC columns are ideal for EPA methods 624 and 608.
  • Mid-polarity phase
  • Allows resolution of 2-nitropropane from 1,1-dichloropropanone under EPA Method 524.2 revision IV
Order Now
Specificaties
Environmentally Preferable