Blood collecting system, S-Monovette® (Fluoride)
Leverancier: SARSTEDT
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
04.1903.001 04.1903 05.1073 05.1073.001 06.1665.001
SARS04.1903.001EA 99.8 EUR
SARS04.1903.001 SARS04.1903 SARS05.1073 SARS05.1073.001 SARS06.1665.001
Blood collecting system, S-Monovette® (Fluoride)
Diagnostische systemen Clinical Sample Collection/Preparation Blood Sample Collection/Preparation
Blood collecting system for the glucose determination (fluoride).