Blood collecting system, S-Monovette®, ESR/Citrate buffer
Leverancier: SARSTEDT
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
05.1079 05.1079.001
SARS05.1079EA 99.6 EUR
SARS05.1079 SARS05.1079.001
Blood collecting system, S-Monovette®, ESR/Citrate buffer
Diagnostische systemen Clinical Sample Collection/Preparation Blood Sample Collection/Preparation
This blood collection system enables hygienic and easy handling in an enclosed system.
Minimal patient discomfort when collecting blood
Higher performance than Westergren method
HD-PE piston with PS plunger