Blood collecting system, S-Monovette® LH (on beads)
Leverancier: SARSTEDT
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
02.1065.001 05.1553.001 01.1604 03.1628.001 05.1106 05.1553 01.1604.001 02.1065 03.1628 04.1906 04.1906.001-50 04.1936 04.1936.001 05.1106.001 06.1666.001
552-5716EA 147 EUR
552-5716 SARS05.1553.001 SARS01.1604 SARS03.1628.001 SARS05.1106 SARS05.1553 SARS01.1604.001 SARS02.1065 SARS03.1628 SARS04.1906 SARS04.1906.001-50 SARS04.1936 SARS04.1936.001 SARS05.1106.001 SARS06.1666.001
Blood collecting system, S-Monovette® LH (on beads)
Diagnostische systemen Clinical Sample Collection/Preparation Blood Sample Collection/Preparation
This blood collection system enables hygienic and easy handling in an enclosed system.
Minimal patient discomfort when collecting blood
Higher performance than Westergren method
HD-PE piston with PS plunger
Order Now
Specificaties
Environmentally Preferable