Capillary columns, TraceGOLD™ TG-5SilMS
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
26096-2230 26096-1300 26096-1420 26096-1430 26096-1540 26096-2240 26096-2960 26096-2970 26096-3080 26096-3360
555-8658EA 901 EUR
555-8658 555-8667 555-8664 555-8661 555-8660 555-8656 555-8650 555-8649 555-8648 555-8647
Capillary columns, TraceGOLD™ TG-5SilMS
Kolommen voor chromatografie
These columns incorporate phenyl groups in the polymer backbone for improved thermal stability, reduced bleed and reduced susceptibility to oxidation.
  • Designed for low bleed and outstanding inertness
  • Low polarity, silarylene phase with selectivity comparable to 5% diphenyl or 95% dimethyl polysiloxane