Capillary columns, TraceGOLD™ TG-VRX
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
26081-3320 26081-3330 26081-3410 26081-4950 26081-4960 26081-3390
555-9004EA 881 EUR
555-9004 555-9003 555-9001 555-8996 555-8995 555-9002
Capillary columns, TraceGOLD™ TG-VRX
Kolommen voor chromatografie
TraceGOLD™ TG-VRX GC columns are ideal for volatile organic pollutants.
  • Highly stable polymer phase
  • Low bleed for excellent signal-to-noise ratio, sensitivity and mass spectral integrity