Capillary columns, TracePLOT™ TG-BOND Alumina
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
26001-6080 26001-6050 26001-6110 26002-6020 26002-6050 26002-6080 26002-6110 26001-6020
555-8829EA 1060 EUR
555-8829 555-8830 555-8828 555-8827 555-8826 555-8879 555-8878 555-8831
Capillary columns, TracePLOT™ TG-BOND Alumina
Kolommen voor chromatografie
Optimised for linear and quantitative analysis of polar unsaturated hydrocarbons.
  • Strong bonding to prevent particle generation
  • Columns to which water has adsorbed may be regenerated to restore full efficiency and selectivity
Each column has been tested to ensure proper film thickness (1,3-butadiene), selectivity (propadiene and methyl acetylene), resolution (trans-2-butene and 1-butene) and coating efficiency (1,3-butadiene).