Magnetic stirrer, accuMIX
Leverancier: 2MAG
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
30401_UK 30401_CH
TMAG30401_UKEA 635 EUR
TMAG30401_UK TMAG30401_CH
Magnetic stirrer, accuMIX
Roerders Magnetische roerders
Ultra flat, wear-free magnetic stirrer with stainless steel housing constructed for the independent battery- and stationary mains operation.
  • Maintenance- and wear-free inductive drive
  • Jerk-free stirring at low speeds
  • Two-step power setting for high power with regard to large and viscous stirring amounts
  • Reduced power for battery-saving long-term operation
SoftStart for reliable catching/centering and safe accelerating of stirring bar.
Certificaten: IP protection class: IP 30
Leveringsinformatie: Supplied with power supply unit, 12 V car adapterMore Product Information
Order Now
Specificaties
Model accuMIX
Environmentally Preferable
Toerentalbereik 100 - 1600 min⁻¹
Max. roercapaciteit H₂O 1 - 3000 ml
Roervermogen 2 or 10 W
Plaatmateriaal Roestvrij staal
Gewicht 2,6 kg
b×d×h 215×130×40 mm