HPLC columns, BioBasic™
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
72105-014001 72105-011001 72305-014001 72205-254630 72105-254630 72105-152130 72105-102130 72105-052130 72305-052130 72105-012101 72305-254630 72305-154630 72105-151030 72305-051030 72305-012101 72305-104630 72305-102130 72305-101030 72305-252130 72305-154030 72305-152130 72105-252130 72105-051030 72105-101030 72105-154630
HYPE72105-014001EA 517 EUR
HYPE72105-014001 HYPE72105-011001 HYPE72305-014001 HYPE72205-254630 HYPE72105-254630 HYPE72105-152130 HYPE72105-102130 HYPE72105-052130 HYPE72305-052130 HYPE72105-012101 HYPE72305-254630 HYPE72305-154630 HYPE72105-151030 HYPE72305-051030 HYPE72305-012101 HYPE72305-104630 HYPE72305-102130 HYPE72305-101030 HYPE72305-252130 HYPE72305-154030 HYPE72305-152130 HYPE72105-252130 HYPE72105-051030 HYPE72105-101030 HYPE72105-154630
HPLC columns, BioBasic™
Kolommen voor chromatografie
The BioBasic™ HPLC columns are ideal for analysis of proteins, peptides and other biomolecules. The BioBasic reversed phase packings are available in C18, C8, C4, phenyl and cyano chemistries.