Intensifying screens, BioMax® TranScreen HE
Leverancier: Kodak
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Kodak® BioMax®
730-3246EA 0 EUR
730-3246 730-3235 KODK8041196 KODK8356800
Intensifying screens, BioMax® TranScreen HE
Autoradiografiefilms Folies
Designed for use with gamma emitters such as ¹²⁵I and high-energy beta emitters, such as ³²P, the BioMax™ TranScreen HE intensifying screen offers the greatest sensitivity. When used in conjunction with the BioMax™ MS film, the system equals or exceeds the image quality of conventional autoradiography screen/film systems. By minimising the distances between the radioisotope and phosphor layer, it presents the phosphor to the emitting particles immediately, for less scattering and better spatial resolution.