Intensifying screens, BioMax® MS
Leverancier: CARESTREAM HEALTH
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
8981706 8770331
KODK8981706EA 0 EUR
KODK8981706 730-2542
Intensifying screens, BioMax® MS
Autoradiografiefilms Folies
The BioMax® MS intensifying screen is optimised for use with BioMax® MS film to maximise sensitivity for detection of ³²P and ¹²⁵I labelled samples.
The BioMax® MS intensifying screen may be used with any autoradiography film to increase sensitivity. However, maximum sensitivity is achieved in combination with the BioMax® MS film.